missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,6-Dichloro-3-methylpyridine, 98%

Manufacturer:  Alfa Aesar™ H6127103

 View more versions of this product

Catalog No. AAH6127103



Description

Description

Applications
2,6-Dichloro-3-methylpyridine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Slightly soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications

Specifications

2,6-Dichloro-3-methylpyridine
58584-94-4
MFCD00137847
Slightly soluble in water.
CC1=C(N=C(C=C1)Cl)Cl
162.013
162.02
98%
1g
C6H5Cl2N
2,6-bis chloranyl-3-methyl-pyridine
KNIKCOPEHZTQJV-UHFFFAOYSA-N
2,6-dichloro-3-methylpyridine
817088
Solid

Chemical Identifiers

58584-94-4
162.013
KNIKCOPEHZTQJV-UHFFFAOYSA-N
817088
CC1=C(N=C(C=C1)Cl)Cl
C6H5Cl2N
MFCD00137847
2,6-bis chloranyl-3-methyl-pyridine
2,6-dichloro-3-methylpyridine
Safety and Handling

Safety and Handling

GHS H Statement
H301-H318-H315-H335
Toxic if swallowed.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c

H302-H315-H318-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents