Learn More
2,6-Dichloro-3-iodopyridine, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals H6436606
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsIt is used as pharmaceutical raw material and intermediate.
Solubility
Slightly soluble in water.
Notes
Light Sensitive. Store in dark. Store away from oxidizing agent.
Specifications
2,6-Dichloro-3-iodopyridine | |
74°C to 79°C | |
C5H2Cl2IN | |
5 g | |
6135696 | |
2,6-dichloro-3-iodo-pyridine, pyridine, 2,6-dichloro-3-iodo, zlchem 993, acmc-20aihk, pubchem14239, tpc-py080, 2,6-dichloro-3-iodo pyridine, pyridine,2,6-dichloro-3-iodo, 2,6-dichloro-3-iodopyridine | |
DPCQIHCGMIPSQV-UHFFFAOYSA-N | |
2,6-dichloro-3-iodopyridine | |
2736002 | |
98% |
148493-37-2 | |
White to Brown | |
MFCD03094686 | |
UN2811 | |
Light sensitive | |
Slightly soluble in water. | |
C1=CC(=NC(=C1I)Cl)Cl | |
273.882 | |
273.88 | |
Powder |
Chemical Identifiers
148493-37-2 | |
273.882 | |
DPCQIHCGMIPSQV-UHFFFAOYSA-N | |
2,6-dichloro-3-iodopyridine |
C5H2Cl2IN | |
MFCD03094686 | |
2736002 | |
C1=CC(=NC(=C1I)Cl)Cl |
Safety and Handling
GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold