Please login to your online account to display your discounted pricing

Alfa Aesar™ 2',6'-Dichloro-3'-fluoroacetophenone, 98%

$62.96 - $187.80

Chemical Identifiers

Chemical Name or Material 2',6'-Dichloro-3'-fluoroacetophenone
CAS 290835-85-7
Molecular Formula C8H5Cl2FO
Formula Weight 207.03
MDL Number MFCD02093760
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2663806

  SDS
alfa aesar™
H2663806
5g Each for $62.96

AAH2663814

  SDS
alfa aesar™
H2663814
25g Each for $187.80
Description & Specifications

Specifications

Formula Weight 207.03
Chemical Name or Material 2',6'-Dichloro-3'-fluoroacetophenone
Assay 98%
Molecular Formula C8H5Cl2FO
MDL Number MFCD02093760
CAS 290835-85-7
Boiling Point 255°C
Flash Point >110°C (230°F)
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.