Please login to your online account to display your discounted pricing

2,6-dichloro-1,3-benzothiazole, Acros Organics

Chemical Identifiers

Chemical Name or Material 2, 6-Dichloro-1, 3-benzothiazole
Assay 97%
Assay Percent Range 97%
Molecular Formula C7H3Cl2NS
Formula Weight 204.08
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SB01025EA

  SDS
fisher scientific company
SB01025EA
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 204.08
Quantity 10g
Assay 97%
Assay Percent Range 97%
Molecular Formula C7H3Cl2NS
Chemical Name or Material 2, 6-Dichloro-1, 3-benzothiazole
CAS 3622-23-9