Please login to your online account to display your discounted pricing

2,6-dichloro-1,3-benzothiazole, Acros Organics


Quantity 10g

Manufacturer: fisher scientific company  SB01025EA

 View more versions of this product

Catalog No. SB01025EA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 204.08
Quantity 10g
Assay 97%
Chemical Name or Material 2, 6-Dichloro-1, 3-benzothiazole
Assay Percent Range 97%
Molecular Formula C7H3Cl2NS
CAS 3622-23-9

Specifications

Chemical Name or Material 2, 6-Dichloro-1, 3-benzothiazole
Assay 97%
Assay Percent Range 97%
Molecular Formula C7H3Cl2NS
Formula Weight 204.08