missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,6-Dibromotoluene, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1183214

 View more versions of this product

Catalog No. AAA1183214



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2,6-Dibromotoluene
2°C to 6°C
112°C to 113°C (7 mmHg)
1.606
25 g
2,6-dibromotoluene, benzene, 1,3-dibromo-2-methyl, 1,3-dibromo-2-methyl-benzene, pubchem3926, acmc-1bagq, ksc493g1t, #, 2,6-dibromotoluene 1g
CC1=C(Br)C=CC=C1Br
249.93
249.94
69321-60-4
1.812
C7H6Br2
MFCD00013524
3235502
OCSKCBIGEMSDIS-UHFFFAOYSA-N
1,3-dibromo-2-methylbenzene
34681
≥98%

Chemical Identifiers

69321-60-4
249.93
OCSKCBIGEMSDIS-UHFFFAOYSA-N
34681
CC1=C(Br)C=CC=C1Br
C7H6Br2
MFCD00013524
2,6-dibromotoluene, benzene, 1,3-dibromo-2-methyl, 1,3-dibromo-2-methyl-benzene, pubchem3926, acmc-1bagq, ksc493g1t, #, 2,6-dibromotoluene 1g
1,3-dibromo-2-methylbenzene
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only