missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,6-Dibromopyridine, 98%

Manufacturer:  Alfa Aesar™ A1539714*

 View more versions of this product

Catalog No. AAA1539714



Description

Description

Applications
2,6-Dibromopyridine is used as a tridentate chelating ligand and in the formation of macrocycles containing the terpyridine moiety. It is also used to produce 6-bromo-2-methoxypyridine.

Solubility
Soluble in hot methanol. Insoluble in water.

Notes
Incompatible with oxidizing agents. Store in a cool place.
Specifications

Specifications

2,6-Dibromopyridine
Recognizable
117°C to 120°C
626-05-1
MFCD00006223
108922
FEYDZHNIIMENOB-UHFFFAOYSA-N
2,6-dibromopyridine
12274
98%
213°C (415°F)
255°C
25g
C5H3Br2N
UN2811
pyridine, 2,6-dibromo, 2,6-dibromo-pyridine, 2,6-dibromo pyridine, 2,6dibromopyridine, 2,6-dibrompyridine, 2,6 dibromopyridine, 2.6-dibromopyridine, pubchem3204, 2, 6-dibromopyridine, acmc-209n5v
C1=CC(=NC(=C1)Br)Br
236.894
236.9

Chemical Identifiers

626-05-1
236.894
FEYDZHNIIMENOB-UHFFFAOYSA-N
12274
C1=CC(=NC(=C1)Br)Br
C5H3Br2N
MFCD00006223
pyridine, 2,6-dibromo, 2,6-dibromo-pyridine, 2,6-dibromo pyridine, 2,6dibromopyridine, 2,6-dibrompyridine, 2,6 dibromopyridine, 2.6-dibromopyridine, pubchem3204, 2, 6-dibromopyridine, acmc-209n5v
2,6-dibromopyridine
Safety and Handling

Safety and Handling

GHS H Statement
H300-H312-H332-H315-H319
Fatal if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

RTECSNumber : US7883000

Documents