missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,6-Dibromophenol, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1057306

 View more versions of this product

Catalog No. AAA1057306



Description

Description

2,6-Dibromophenol is used in the production of 3,4,5-trimethoxybenzaldehyde and 2-(2,6-dibromophenoxy)ethylbromide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Dibromophenol is used in the production of 3,4,5-trimethoxybenzaldehyde and 2-(2,6-dibromophenoxy)ethylbromide.

Solubility
Soluble in ethanol and ether. Slightly soluble in water.

Notes
Incompatible with acid chlorides, acid anhydrides and oxidizing agents.
Specifications

Specifications

2,6-Dibromophenol
53°C to 57°C
Phenol-like
MFCD00002152
2043614
Soluble in ethanol and ether. Slightly soluble in water.
OC1=C(Br)C=CC=C1Br
251.91
CHEBI:19391
99%
608-33-3
255°C to 256°C
C6H4Br2O
5 g
phenol, 2,6-dibromo, 2,6-dibromo-phenol, unii-27mip05eav, 2,6-dibromo-pheno, 27mip05eav, 2,6-bis bromanyl phenol, acmc-1au9i, 2,6-dibromophenol, ksc493e5t, 2,6-dibromophenol, analytical standard
SSIZLKDLDKIHEV-UHFFFAOYSA-N
2,6-dibromophenol
11847
251.92

Chemical Identifiers

608-33-3
251.91
SSIZLKDLDKIHEV-UHFFFAOYSA-N
11847
2,6-dibromophenol
C6H4Br2O
MFCD00002152
phenol, 2,6-dibromo, 2,6-dibromo-phenol, unii-27mip05eav, 2,6-dibromo-pheno, 27mip05eav, 2,6-bis bromanyl phenol, acmc-1au9i, 2,6-dibromophenol, ksc493e5t, 2,6-dibromophenol, analytical standard
CHEBI:19391
OC1=C(Br)C=CC=C1Br
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

DOTInformation : Hazard Class: 6.1; Packaging Group: III

EINECSNumber : 210-161-0

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only