missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,6-Dibromophenol 98%, ACROS Organics™

Chemical Identifiers

CAS 608-33-3
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Molecular Formula C6 H4 Br2 O
Linear Formula Br2C6H3OH
Formula Weight 251.91
MDL Number MFCD00002152
Chemical Name or Material 2, 6-Dibromophenol, 98%
Assay ≥98.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC406520010 SDS Thermo Fisher Scientific
40652-0010
Glass bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

608-33-3
100.0
C6 H4 Br2 O
251.91
2, 6-Dibromophenol, 98%
98.5
99%
Br2C6H3OH
MFCD00002152
≥98.5%

Specifications

Crystals or Needles
1g
53° to 57°C
98.5
Glass bottle
608-33-3
MFCD00002152
2, 6-Dibromophenol, 98%
251.91
White
100.0
Authentic
99%
Br2C6H3OH
C6 H4 Br2 O
≥98.5%

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 210-161-0

TSCA : TSCA