missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,6-Dibromobenzo[1,2-b:4,5-b']dithiophene 98.0+%, TCI America™
$406.46 - $1418.44
Chemical Identifiers
CAS | 909280-97-3 |
---|---|
Molecular Formula | C10H4Br2S2 |
Molecular Weight (g/mol) | 348.07 |
InChI Key | SXRSYJAHJIIXFM-UHFFFAOYSA-N |
PubChem CID | 56973692 |
IUPAC Name | 2,6-dibromothieno[2,3-f][1]benzothiole |
SMILES | C1=C2C=C(SC2=CC3=C1SC(=C3)Br)Br |
Chemical Identifiers
909280-97-3 | |
348.07 | |
56973692 | |
C1=C2C=C(SC2=CC3=C1SC(=C3)Br)Br |
C10H4Br2S2 | |
SXRSYJAHJIIXFM-UHFFFAOYSA-N | |
2,6-dibromothieno[2,3-f][1]benzothiole |
Specifications
909280-97-3 | |
Yellow | |
1 g | |
C1=C2C=C(SC2=CC3=C1SC(=C3)Br)Br | |
348.07 | |
348.07 | |
Crystalline Powder |
258°C | |
C10H4Br2S2 | |
SXRSYJAHJIIXFM-UHFFFAOYSA-N | |
2,6-dibromothieno[2,3-f][1]benzothiole | |
56973692 | |
≥98.0% (GC) | |
2,6-Dibromobenzo[1,2-b:4,5-b′]dithiophene |
Safety and Handling
TSCA : No