missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,6-Dibromoaniline, 97%, Acros Organics
Manufacturer: Thermo Fisher Scientific 112700100
Specifications
2, 6-Dibromoaniline | |
Glass bottle | |
Gray or White to Yellow | |
10g | |
608-30-0 | |
100.0 | |
C6H5Br2N | |
12, 659 | |
XIRRDAWDNHRRLB-UHFFFAOYSA-N | |
2,6-dibromoaniline | |
69098 | |
Crystalline Powder |
Authentic | |
262.0°C to 264.0°C | |
79.0°C to 84.0°C | |
97% | |
96.0 | |
96% min. (GC) | |
Br2C6H3NH2 | |
2,6-dibromo-aniline | |
C1=CC(=C(C(=C1)Br)N)Br | |
250.92 | |
250.92 | |
97% |
Chemical Identifiers
608-30-0 | |
250.92 | |
2,6-dibromo-aniline | |
2,6-dibromoaniline |
C6H5Br2N | |
XIRRDAWDNHRRLB-UHFFFAOYSA-N | |
69098 | |
C1=CC(=C(C(=C1)Br)N)Br |
Safety and Handling
GHS H Statement
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
GHS Signal Word: Warning
EINECSNumber : 210-158-4
TSCA : TSCA