missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,6-Dibromo-N-chloro-p-benzoquinoneimine, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 406470050

 View more versions of this product

Catalog No. AC406470050


 

Specifications Description & Specifications

Specifications

2, 6-Dibromo-N-chloro-p-benzoquinoneimine
Yellow to ochre
5g
Authentic
100.0
Product may discolor to orange-brown on storage
≥96.0%
C6H2Br2ClNO
2, 6-Dibromoquinonechloroimide
1.8100g/mL
76° to 83°C
537-45-1
96.0
Glass bottle
Powder or Fine Crystalline Needles
97%
299.34

Chemical Identifiers

537-45-1
100.0
C6H2Br2ClNO
2, 6-Dibromoquinonechloroimide
≥96.0%
96.0
97%
299.34
2, 6-Dibromo-N-chloro-p-benzoquinoneimine

Safety and Handling

SDS & Safety Documents

GHS P Statement
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Harmful in contact with skin.
Harmful if inhaled.
Heating may cause a fire.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger