Please login to your online account to display your discounted pricing

2,6-Dibromo-4-(trifluoromethyl)aniline, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 2, 6-Dibromo-4-(trifluoromethyl)aniline
CAS 72678-19-4
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C7H4Br2F3N
Formula Weight 318.92
MDL Number MFCD00068181
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SEW02789DA

 
fisher scientific company
SEW02789DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 318.92
Quantity 1g
Assay 97%
CAS Min % 97.0
Packaging Amber Glass Bottle
CAS 72678-19-4
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 2, 6-Dibromo-4-(trifluoromethyl)aniline
Assay Percent Range 97%
Molecular Formula C7H4Br2F3N
MDL Number MFCD00068181
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.