missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,6-Dibromo-4-(trifluoromethyl)aniline, 97%

Manufacturer:  Alfa Aesar™ L1065806

 View more versions of this product

Catalog No. AAL1065806



Description

Description

Applications
Used in enantio-and diastereoselective addition reation.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Specifications

2,6-Dibromo-4-(trifluoromethyl)aniline
36°C to 40°C
72678-19-4
MFCD00068181
6314196
DRSMEHXBOXHXDX-UHFFFAOYSA-N
2,6-dibromo-4-(trifluoromethyl)aniline
144565
97%
64°C to 65°C (0.1mmHg)
5g
C7H4Br2F3N
UN2811
2,6-dibromo-4-trifluoromethyl aniline, 4-amino-3,5-dibromobenzotrifluoride, 2,6-dibromo-4-trifluoromethyl phenylamine, benzenamine,2,6-dibromo-4-trifluoromethyl, benzenamine, 2,6-dibromo-4-trifluoromethyl, pubchem2805, acmc-209ony, buttpark 29\06-99, 2,6-dibromo-4-trifluoromethyl aniline, 2,6-dibromo-4-trifluoromethyl-phenylamine
C1=C(C=C(C(=C1Br)N)Br)C(F)(F)F
318.919
318.93

Chemical Identifiers

72678-19-4
318.919
DRSMEHXBOXHXDX-UHFFFAOYSA-N
144565
C1=C(C=C(C(=C1Br)N)Br)C(F)(F)F
C7H4Br2F3N
MFCD00068181
2,6-dibromo-4-trifluoromethyl aniline, 4-amino-3,5-dibromobenzotrifluoride, 2,6-dibromo-4-trifluoromethyl phenylamine, benzenamine,2,6-dibromo-4-trifluoromethyl, benzenamine, 2,6-dibromo-4-trifluoromethyl, pubchem2805, acmc-209ony, buttpark 29\06-99, 2,6-dibromo-4-trifluoromethyl aniline, 2,6-dibromo-4-trifluoromethyl-phenylamine
2,6-dibromo-4-(trifluoromethyl)aniline
Safety and Handling

Safety and Handling

GHS H Statement
H311-H302-H332-H315-H319-H335
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

Documents