missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,6-Dibromo-4-(trifluoromethyl)aniline, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L1065803

 View more versions of this product

Catalog No. AAL1065803



Description

Description

Used in enantio-and diastereoselective addition reation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in enantio-and diastereoselective addition reation.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Specifications

2,6-Dibromo-4-(trifluoromethyl)aniline
36°C to 40°C
C7H4Br2F3N
1 g
6314196
Insoluble in water.
C1=C(C=C(C(=C1Br)N)Br)C(F)(F)F
318.919
318.93
72678-19-4
64°C to 65°C (0.1 mmHg)
MFCD00068181
UN2811
2,6-dibromo-4-trifluoromethyl aniline, 4-amino-3,5-dibromobenzotrifluoride, 2,6-dibromo-4-trifluoromethyl phenylamine, benzenamine,2,6-dibromo-4-trifluoromethyl, benzenamine, 2,6-dibromo-4-trifluoromethyl, pubchem2805, acmc-209ony, buttpark 29\06-99, 2,6-dibromo-4-trifluoromethyl-phenylamine
DRSMEHXBOXHXDX-UHFFFAOYSA-N
2,6-dibromo-4-(trifluoromethyl)aniline
144565
97%

Chemical Identifiers

72678-19-4
318.919
DRSMEHXBOXHXDX-UHFFFAOYSA-N
144565
C1=C(C=C(C(=C1Br)N)Br)C(F)(F)F
C7H4Br2F3N
MFCD00068181
2,6-dibromo-4-trifluoromethyl aniline, 4-amino-3,5-dibromobenzotrifluoride, 2,6-dibromo-4-trifluoromethyl phenylamine, benzenamine,2,6-dibromo-4-trifluoromethyl, benzenamine, 2,6-dibromo-4-trifluoromethyl, pubchem2805, acmc-209ony, buttpark 29\06-99, 2,6-dibromo-4-trifluoromethyl-phenylamine
2,6-dibromo-4-(trifluoromethyl)aniline
Safety and Handling

Safety and Handling

GHS H Statement
H311-H302-H332-H315-H319-H335
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only