missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,6-Dibromo-4-(trifluoromethoxy)aniline 98.0+%, TCI America™
$73.00 - $233.92
Chemical Identifiers
CAS | 88149-49-9 |
---|---|
Molecular Formula | C7H4Br2F3NO |
Molecular Weight (g/mol) | 334.92 |
MDL Number | MFCD00153113 |
InChI Key | JBSWOEGXMADXOU-UHFFFAOYSA-N |
Synonym | 2,6-dibromo-4-trifluoromethoxy aniline, bromo riluzole impurity, 3,5-dibromo-4-aminotrifluoromethoxybenzene, 2,6-dibromo-4-trifluoromethoxy benzenamine, 2-amino-1,3-dibromo-5-trifluoromethoxy benzene, 2,6dibromo-4-trifluoromethoxy aniline, 2,6-dibromo-4-trifluoro-methoxy aniline, benzenamine, 2,6-dibromo-4-trifluoromethoxy, pubchem10031 |
PubChem CID | 2736797 |
IUPAC Name | 2,6-dibromo-4-(trifluoromethoxy)aniline |
SMILES | NC1=C(Br)C=C(OC(F)(F)F)C=C1Br |
Chemical Identifiers
88149-49-9 | |
334.92 | |
JBSWOEGXMADXOU-UHFFFAOYSA-N | |
2736797 | |
NC1=C(Br)C=C(OC(F)(F)F)C=C1Br |
C7H4Br2F3NO | |
MFCD00153113 | |
2,6-dibromo-4-trifluoromethoxy aniline, bromo riluzole impurity, 3,5-dibromo-4-aminotrifluoromethoxybenzene, 2,6-dibromo-4-trifluoromethoxy benzenamine, 2-amino-1,3-dibromo-5-trifluoromethoxy benzene, 2,6dibromo-4-trifluoromethoxy aniline, 2,6-dibromo-4-trifluoro-methoxy aniline, benzenamine, 2,6-dibromo-4-trifluoromethoxy, pubchem10031 | |
2,6-dibromo-4-(trifluoromethoxy)aniline |
Specifications
88149-49-9 | |
C7H4Br2F3NO | |
5 g | |
JBSWOEGXMADXOU-UHFFFAOYSA-N | |
2,6-dibromo-4-(trifluoromethoxy)aniline | |
2736797 | |
≥98.0% (GC) | |
2,6-Dibromo-4-(trifluoromethoxy)aniline |
White-Yellow | |
MFCD00153113 | |
2,6-dibromo-4-trifluoromethoxy aniline, bromo riluzole impurity, 3,5-dibromo-4-aminotrifluoromethoxybenzene, 2,6-dibromo-4-trifluoromethoxy benzenamine, 2-amino-1,3-dibromo-5-trifluoromethoxy benzene, 2,6dibromo-4-trifluoromethoxy aniline, 2,6-dibromo-4-trifluoro-methoxy aniline, benzenamine, 2,6-dibromo-4-trifluoromethoxy, pubchem10031 | |
NC1=C(Br)C=C(OC(F)(F)F)C=C1Br | |
334.92 | |
334.92 | |
Crystalline Powder |
Safety and Handling
EINECSNumber : (3)-4290
TSCA : No