missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,6-Dibromo-4-(trifluoromethoxy)aniline, 97%

Manufacturer:  Alfa Aesar™ H5572503

 View more versions of this product

Catalog No. AAH5572503



Specifications

Specifications

2,6-Dibromo-4-(trifluoromethoxy)aniline
1g
C7H4Br2F3NO
2,6-dibromo-4-trifluoro-methoxy aniline
C1=C(C=C(C(=C1Br)N)Br)OC(F)(F)F
334.918
334.92
70°C to 74°C
88149-49-9
MFCD00153113
JBSWOEGXMADXOU-UHFFFAOYSA-N
2,6-dibromo-4-(trifluoromethoxy)aniline
2736797
97%

Chemical Identifiers

88149-49-9
334.918
JBSWOEGXMADXOU-UHFFFAOYSA-N
2736797
C1=C(C=C(C(=C1Br)N)Br)OC(F)(F)F
C7H4Br2F3NO
MFCD00153113
2,6-dibromo-4-trifluoro-methoxy aniline
2,6-dibromo-4-(trifluoromethoxy)aniline
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents