Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,6-Dibromo-4-nitroaniline, 97+%

$43.73 - $122.38

Chemical Identifiers

Chemical Name or Material 2,6-Dibromo-4-nitroaniline
CAS 827-94-1
Molecular Formula C6H4Br2N2O2
Formula Weight 295.92
MDL Number MFCD00007639
Beilstein 2110915
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1660318

 
alfa aesar™
A1660318
50g Each for $43.73

AAA1660330

 
alfa aesar™
A1660330
250g Each for $122.38
Description & Specifications

Specifications

Formula Weight 295.92
MDL Number MFCD00007639
Chemical Name or Material 2,6-Dibromo-4-nitroaniline
TSCA TSCA
Melting Point 205° to 208°C
Assay ≥97%
Quantity 50g
Molecular Formula C6H4Br2N2O2
CAS 827-94-1
Beilstein 2110915
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H311-H373-H302-H332-H315-H319-H335
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P260-P261-P305+P351+P338-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number BX2599000
Safety and Handling

Danger

GHS P Statement
P260-P261-P305+P351+P338-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H311-H373-H302-H332-H315-H319-H335
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.