Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,6-Dibromo-4-n-propylaniline, 98%

$53.49 - $628.32

Chemical Identifiers

Chemical Name or Material 2,6-Dibromo-4-n-propylaniline
CAS 10546-64-2
Molecular Formula C9H11Br2N
Formula Weight 293.01
MDL Number MFCD00051706
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2480206

  SDS
alfa aesar™
B2480206
5g Each for $53.49

AAB2480214

  SDS
alfa aesar™
B2480214
25g Each for $197.39

AAB2480222

  SDS
alfa aesar™
B2480222
100g Each for $628.32
Description & Specifications

Specifications

Formula Weight 293.01
Chemical Name or Material 2,6-Dibromo-4-n-propylaniline
Melting Point 55° to 57°C
Assay 98%
Quantity 5g
Molecular Formula C9H11Br2N
MDL Number MFCD00051706
CAS 10546-64-2
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.