Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,6-Dibromo-4-methylphenol, 98%

$64.50 - $64.50

Chemical Identifiers

Chemical Name or Material 2,6-Dibromo-4-methylphenol
CAS 2432-14-6
Molecular Formula C7H6Br2O
Formula Weight 265.94
MDL Number MFCD00002154
Beilstein 2045283
Synonym 2,6-Dibromo-p-cresol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1863414

  SDS
alfa aesar™
A1863414
25g Each for $64.50
Description & Specifications

Specifications

Formula Weight 265.94
Chemical Name or Material 2,6-Dibromo-4-methylphenol
Melting Point 49° to 50°C
Assay 98%
Quantity 25g
Flash Point >110°C (230°F)
Molecular Formula C7H6Br2O
MDL Number MFCD00002154
CAS 2432-14-6
Beilstein 2045283
Synonym 2,6-Dibromo-p-cresol
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number GO7700000
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.