Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,6-Dibromo-4-fluorophenyl isocyanate, tech. 90%

Chemical Identifiers

Chemical Name or Material 2,6-Dibromo-4-fluorophenyl isocyanate
CAS 76393-18-5
Purity Grade Technical Grade
Molecular Formula C7H2Br2FNO
Formula Weight 294.91
MDL Number MFCD00037041
UN Number UN2811
Beilstein 8617287
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1874403

 
alfa aesar™
L1874403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1874406

 
alfa aesar™
L1874406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 294.91
Chemical Name or Material 2,6-Dibromo-4-fluorophenyl isocyanate
Assay 90%
Molecular Formula C7H2Br2FNO
MDL Number MFCD00037041
CAS 76393-18-5
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H331-H334-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P284a-P261-P280-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Purity Grade Technical Grade
Melting Point 42° to 45°C
Quantity 1g
Flash Point >110°C (230°F)
UN Number UN2811
Beilstein 8617287
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P284a-P261-P280-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H334-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.