Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,6-Dibromo-4-fluorophenol, 98+%

$1,002.00 - $1,002.00

Chemical Identifiers

Chemical Name or Material 2,6-Dibromo-4-fluorophenol
CAS 344-20-7
Molecular Formula C6H3Br2FO
Formula Weight 269.91
MDL Number MFCD00002317
Beilstein 1868033
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2478222

  SDS
alfa aesar™
B2478222
100g Each for $1,002.00
Description & Specifications

Specifications

Formula Weight 269.91
Chemical Name or Material 2,6-Dibromo-4-fluorophenol
Melting Point 54° to 56°C
Assay ≥98%
Quantity 100g
Molecular Formula C6H3Br2FO
Odor Phenol-like
MDL Number MFCD00002317
CAS 344-20-7
Beilstein 1868033
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.