Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,6-Dibromo-4-chloroaniline, 98+%

Chemical Identifiers

Chemical Name or Material 2,6-Dibromo-4-chloroaniline
CAS 874-17-9
Molecular Formula C6H4Br2ClN
Formula Weight 285.38
MDL Number MFCD00051750
Beilstein 2803730
Synonym 4-Chloro-2,6-dibromoaniline
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1913418

 
alfa aesar™
A1913418
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material 2,6-Dibromo-4-chloroaniline
Melting Point 91° to 95°C
Quantity 50g
CAS 874-17-9
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay ≥98%
Molecular Formula C6H4Br2ClN
Formula Weight 285.38
MDL Number MFCD00051750
Beilstein 2803730
Synonym 4-Chloro-2,6-dibromoaniline