Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,6-Dibromo-4-chloro-3,5-dimethylphenol, 97%

$355.99 - $355.99

Chemical Identifiers

Chemical Name or Material 2,6-Dibromo-4-chloro-3,5-dimethylphenol
CAS 175204-32-7
Molecular Formula C8H7Br2ClO
Formula Weight 314.40g/mol
MDL Number MFCD00219924
Synonym 4-Chloro-2,6-dibromo-3,5-dimethylphenol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2518406

 
alfa aesar™
B25184-06
5g Each for $355.99
Description & Specifications

Specifications

Melting Point 158° to 160°C
Quantity 5g
Chemical Name or Material 2,6-Dibromo-4-chloro-3,5-dimethylphenol
CAS 175204-32-7
Assay 97%
Molecular Formula C8H7Br2ClO
Formula Weight 314.40g/mol
MDL Number MFCD00219924
Synonym 4-Chloro-2,6-dibromo-3,5-dimethylphenol
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.