Please login to your online account to display your discounted pricing

Alfa Aesar™ 2',6-Dibromo-3,4'-bipyridine, 95%

$174.78 - $622.30

Chemical Identifiers

Chemical Name or Material 2',6-Dibromo-3,4'-bipyridine
CAS 942206-16-8
Molecular Formula C10H6Br2N2
Formula Weight 313.98
MDL Number MFCD09037424
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH27214MC

  SDS
alfa aesar™
H27214MC
100mg Each for $174.78

AAH27214ME

  SDS
alfa aesar™
H27214ME
500mg Each for $622.30
Description & Specifications

Specifications

Formula Weight 313.98
Chemical Name or Material 2',6-Dibromo-3,4'-bipyridine
Health Hazard 1 Warning
Quantity 100mg
Assay 95%
MDL Number MFCD09037424
CAS 942206-16-8
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Melting Point 189° to 193°C
Molecular Formula C10H6Br2N2
Safety and Handling

Warning

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.