missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,6-Diaminopyridine, 98%, Thermo Scientific™

Manufacturer:  Thermo Fisher Scientific 112381000

 View more versions of this product

Catalog No. AC112381000



Description

Description

Specifications

Specifications

2, 6-Diaminopyridine
Authentic
1% max.
Beige to Brown-Gray
100g
141-86-6
100.0
C5H7N3
MFCD00006329
2,6-diamino pyridine, 2,6-diaminopyridine, 2,6-pyridinediamine, ccris 6682, dap amine, dsstox_cid_20127, k89ab8ckg6, pyridine, 2,6-diamino, pyridine-2,6-diyldiamine, unii-k89ab8ckg6
VHNQIURBCCNWDN-UHFFFAOYSA-N
pyridine-2,6-diamine
8861
Flakes or Crystalline Powder
155°C
Glass bottle
285°C
115°C to 123°C
98%
97.5
97.5% min. (GC)
(C5H3N)(NH2)2
22,I,647
Solubility in water: 9.9g/100 mL (20°C)
C1=CC(=NC(=C1)N)N
109.13
109.13
98%

Chemical Identifiers

141-86-6
109.13
VHNQIURBCCNWDN-UHFFFAOYSA-N
8861
C1=CC(=NC(=C1)N)N
C5H7N3
MFCD00006329
2,6-diamino pyridine, 2,6-diaminopyridine, 2,6-pyridinediamine, ccris 6682, dap amine, dsstox_cid_20127, k89ab8ckg6, pyridine, 2,6-diamino, pyridine-2,6-diyldiamine, unii-k89ab8ckg6
pyridine-2,6-diamine
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if inhaled.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume

GHS Signal Word: Danger

EINECSNumber : 205-507-2

RTECSNumber : US7570000

TSCA : TSCA

Recommended Storage : Product darkens on storage

SDS Documents
Product Certifications
Promotions

Promotions

RUO – Research Use Only