missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,6-Diaminopyridine, 98%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC112380250
Specifications
2, 6-Diaminopyridine | |
Authentic | |
1% max. | |
Beige to Brown-Gray | |
25g | |
141-86-6 | |
100.0 | |
C5H7N3 | |
MFCD00006329 | |
2,6-diamino pyridine, 2,6-diaminopyridine, 2,6-pyridinediamine, ccris 6682, dap amine, dsstox_cid_20127, k89ab8ckg6, pyridine, 2,6-diamino, pyridine-2,6-diyldiamine, unii-k89ab8ckg6 | |
VHNQIURBCCNWDN-UHFFFAOYSA-N | |
pyridine-2,6-diamine | |
8861 | |
Flakes or Crystalline Powder |
155°C | |
Glass bottle | |
285°C | |
115°C to 123°C | |
98% | |
97.5 | |
97.5% min. (GC) | |
(C5H3N)(NH2)2 | |
22, I, 647 | |
Solubility in water: 9.9g/100 mL (20°C) | |
C1=CC(=NC(=C1)N)N | |
109.13 | |
109.13 | |
98% |
Chemical Identifiers
141-86-6 | |
109.13 | |
VHNQIURBCCNWDN-UHFFFAOYSA-N | |
8861 | |
C1=CC(=NC(=C1)N)N |
C5H7N3 | |
MFCD00006329 | |
2,6-diamino pyridine, 2,6-diaminopyridine, 2,6-pyridinediamine, ccris 6682, dap amine, dsstox_cid_20127, k89ab8ckg6, pyridine, 2,6-diamino, pyridine-2,6-diyldiamine, unii-k89ab8ckg6 | |
pyridine-2,6-diamine |
Safety and Handling
GHS H Statement
Toxic if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if inhaled.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume
GHS Signal Word: Danger
EINECSNumber : 205-507-2
RTECSNumber : US7570000
TSCA : TSCA
Recommended Storage : Product darkens on storage