missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,6-Diaminopurine, 98%

Manufacturer:  Alfa Aesar™ A1564906

 View more versions of this product

Catalog No. AAA1564906



Specifications

Specifications

2,6-Diaminopurine
5g
C5H6N6
9624
2,6-diaminopurine, 9h-purine-2,6-diamine, 2-aminoadenine, 1h-purine-2,6-diamine, 2,6-diamino-9h-purine, purine, 2,6-diamino, purine-2,6-diyldiamine, unii-49p95bau4z, ccris 923, purine-2,6-diamine
C1=NC2=C(N1)C(=NC(=N2)N)N
150.145
CHEBI:40235
98%
~302°C
1904-98-9
MFCD00213668
14,2985
MSSXOMSJDRHRMC-UHFFFAOYSA-N
7H-purine-2,6-diamine
30976
150.14

Chemical Identifiers

1904-98-9
150.145
MSSXOMSJDRHRMC-UHFFFAOYSA-N
30976
7H-purine-2,6-diamine
C5H6N6
MFCD00213668
2,6-diaminopurine, 9h-purine-2,6-diamine, 2-aminoadenine, 1h-purine-2,6-diamine, 2,6-diamino-9h-purine, purine, 2,6-diamino, purine-2,6-diyldiamine, unii-49p95bau4z, ccris 923, purine-2,6-diamine
CHEBI:40235
C1=NC2=C(N1)C(=NC(=N2)N)N
Safety and Handling

Safety and Handling

GHS H Statement
H341
Suspected of causing genetic defects.

GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : UO7524000

TSCA : TSCA

Documents