missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,6-Diaminopimelic acid, 98%, Alfa Aesar

Manufacturer:  Alfa Aesar B2239103

 View more versions of this product

Catalog No. AAB2239103



Description

Description

Applications
2,6-Diaminopimelic acid is used as a constituent of proteoglycans. It is used as a component of bacterial cell wall.

Solubility
Soluble in water, diluted acids and alkali.

Notes
Incompatible with strong oxidizing agents.
Specifications

Specifications

2, 6-Diaminopimelic acid
583-93-7
C7H14N2O4
GMKMEZVLHJARHF-UHFFFAOYSA-N
2,6-diaminoheptanedioic acid
865
190.2
1g
0.98
2,6-diaminopimelic acid, heptanedioic acid, 2,6-diamino, diaminopimelic acid, dl-2,6-diaminoheptanedioic acid, m-dap, dl-alpha,epsilon-diaminopimelic acid, 1,5-diaminoheptanedioic acid, ll-diaminopimelate, d,l-diaminopimelate, ll-a2pm
C(CC(C(=O)O)N)CC(C(=O)O)N
190.199
CHEBI:23673

Chemical Identifiers

583-93-7
190.199
2,6-diaminopimelic acid, heptanedioic acid, 2,6-diamino, diaminopimelic acid, dl-2,6-diaminoheptanedioic acid, m-dap, dl-alpha,epsilon-diaminopimelic acid, 1,5-diaminoheptanedioic acid, ll-diaminopimelate, d,l-diaminopimelate, ll-a2pm
CHEBI:23673
C(CC(C(=O)O)N)CC(C(=O)O)N
C7H14N2O4
GMKMEZVLHJARHF-UHFFFAOYSA-N
865
2,6-diaminoheptanedioic acid
SDS Documents