missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,6-Diaminoheptanedioic acid, 95%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC235540250

 View more versions of this product

Catalog No. AC235540250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

2, 6-Diaminoheptanedioic acid
Glass bottle
300°C
583-93-7
100.0
C7H14N2O4
MFCD00002637
Solubility in water: 0.92% (21°C). Other solubilities: soluble in diluted acids and alkalies
C(CC(C(=O)O)N)CC(C(=O)O)N
190.2
CHEBI:23673
Powder and/or Chunks
Authentic
White to Beige
25g
95.0
95%
HOOCCH(NH2)3(CH2)3CH(NH2)COOH
1,5-diaminoheptanedioic acid, 2,6-diaminopimelic acid, d,l-diaminopimelate, diaminopimelic acid, dl-2,6-diaminoheptanedioic acid, dl-alpha,epsilon-diaminopimelic acid, heptanedioic acid, 2,6-diamino, ll-a2pm, ll-diaminopimelate, m-dap
GMKMEZVLHJARHF-UHFFFAOYSA-N
2,6-diaminoheptanedioic acid
865
190.2
95%

Chemical Identifiers

583-93-7
190.2
GMKMEZVLHJARHF-UHFFFAOYSA-N
865
2,6-diaminoheptanedioic acid
C7H14N2O4
MFCD00002637
1,5-diaminoheptanedioic acid, 2,6-diaminopimelic acid, d,l-diaminopimelate, diaminopimelic acid, dl-2,6-diaminoheptanedioic acid, dl-alpha,epsilon-diaminopimelic acid, heptanedioic acid, 2,6-diamino, ll-a2pm, ll-diaminopimelate, m-dap
CHEBI:23673
C(CC(C(=O)O)N)CC(C(=O)O)N
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning

EINECSNumber : 209-524-6

SDS Documents