missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,6-Diaminoheptanedioic acid, 95%, ACROS Organics™
Manufacturer: Fisher Scientific Acros 235540050
Specifications
2, 6-Diaminoheptanedioic acid | |
Glass bottle | |
300°C | |
583-93-7 | |
100.0 | |
C7H14N2O4 | |
MFCD00002637 | |
Solubility in water: 0.92% (21°C). Other solubilities: soluble in diluted acids and alkalies | |
C(CC(C(=O)O)N)CC(C(=O)O)N | |
190.2 | |
CHEBI:23673 | |
Powder and/or Chunks |
Authentic | |
White to Beige | |
5g | |
95.0 | |
95% | |
HOOCCH(NH2)3(CH2)3CH(NH2)COOH | |
1,5-diaminoheptanedioic acid, 2,6-diaminopimelic acid, d,l-diaminopimelate, diaminopimelic acid, dl-2,6-diaminoheptanedioic acid, dl-alpha,epsilon-diaminopimelic acid, heptanedioic acid, 2,6-diamino, ll-a2pm, ll-diaminopimelate, m-dap | |
GMKMEZVLHJARHF-UHFFFAOYSA-N | |
2,6-diaminoheptanedioic acid | |
865 | |
190.2 | |
95% |
Chemical Identifiers
583-93-7 | |
190.2 | |
GMKMEZVLHJARHF-UHFFFAOYSA-N | |
865 | |
2,6-diaminoheptanedioic acid |
C7H14N2O4 | |
MFCD00002637 | |
1,5-diaminoheptanedioic acid, 2,6-diaminopimelic acid, d,l-diaminopimelate, diaminopimelic acid, dl-2,6-diaminoheptanedioic acid, dl-alpha,epsilon-diaminopimelic acid, heptanedioic acid, 2,6-diamino, ll-a2pm, ll-diaminopimelate, m-dap | |
CHEBI:23673 | |
C(CC(C(=O)O)N)CC(C(=O)O)N |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
EINECSNumber : 209-524-6