Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,6-Diamino-3,5-difluoropyridine, 98%

Chemical Identifiers

Chemical Name or Material 2,6-Diamino-3,5-difluoropyridine
CAS 247069-27-8
Molecular Formula C5H5F2N3
Formula Weight 145.11
MDL Number MFCD03412208
UN Number UN2811
Synonym 3,5-Difluoro-2,6-pyridinediamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3036503

  SDS
alfa aesar™
H3036503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH3036506

  SDS
alfa aesar™
H3036506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 145.11
Chemical Name or Material 2,6-Diamino-3,5-difluoropyridine
Assay 98%
Molecular Formula C5H5F2N3
MDL Number MFCD03412208
CAS 247069-27-8
UN Number UN2811
Synonym 3,5-Difluoro-2,6-pyridinediamine
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H311-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Melting Point 158° to 162°C
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H301-H311-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.