Learn More
2,6-Di-tert-butylpyridine, 97%, Thermo Scientific Chemicals
$299.60 - $299.60
Chemical Identifiers
CAS | 585-48-8 |
---|---|
Molecular Formula | C13H21N |
Molecular Weight (g/mol) | 191.32 |
MDL Number | MFCD00006306 |
InChI Key | UWKQJZCTQGMHKD-UHFFFAOYSA-N |
Synonym | 2,6-di-tert-butylpyridine, 2,6-di tert-butyl pyridine, 2,6-di-t-butylpyridine, unii-oi9lf0h4mm, 2,6-di-t-butyl pyridine, pyridine, 2,6-bis 1,1-dimethylethyl, oi9lf0h4mm, 2,6-di-tert-butyl pyridine, pyridine, 2,6-di-tert-butyl, pubchem16119 |
PubChem CID | 68510 |
IUPAC Name | 2,6-ditert-butylpyridine |
SMILES | CC(C)(C)C1=NC(=CC=C1)C(C)(C)C |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC204640050
|
Thermo Scientific Chemicals
204640050 |
5 g | Glass bottle |
Each for $299.60
|
|
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
585-48-8 | |
191.32 | |
UWKQJZCTQGMHKD-UHFFFAOYSA-N | |
68510 | |
CC(C)(C)C1=NC(=CC=C1)C(C)(C)C |
C13H21N | |
MFCD00006306 | |
2,6-di-tert-butylpyridine, 2,6-di tert-butyl pyridine, 2,6-di-t-butylpyridine, unii-oi9lf0h4mm, 2,6-di-t-butyl pyridine, pyridine, 2,6-bis 1,1-dimethylethyl, oi9lf0h4mm, 2,6-di-tert-butyl pyridine, pyridine, 2,6-di-tert-butyl, pubchem16119 | |
2,6-ditert-butylpyridine |
Specifications
585-48-8 | |
100.0 | |
Yellow | |
208.0°C | |
96% min. (GC) | |
C13H21N | |
MFCD00006306 | |
Glass bottle | |
15, 3045 | |
5 g | |
Solubility in water: insoluble. Other solubilities: soluble in organic solvents | |
UWKQJZCTQGMHKD-UHFFFAOYSA-N | |
2,6-ditert-butylpyridine | |
68510 | |
97% | |
2, 6-Di-tert-butylpyridine, 97% |
96.0 | |
2.0°C to 4.0°C | |
0.8520g/mL | |
72°C | |
Authentic | |
(C5H3N[C(CH3)3]2 | |
20, III, 2868 | |
01,212 | |
1.4720 to 1.4740 | |
2,6-di-tert-butylpyridine, 2,6-di tert-butyl pyridine, 2,6-di-t-butylpyridine, unii-oi9lf0h4mm, 2,6-di-t-butyl pyridine, pyridine, 2,6-bis 1,1-dimethylethyl, oi9lf0h4mm, 2,6-di-tert-butyl pyridine, pyridine, 2,6-di-tert-butyl, pubchem16119 | |
0.852 | |
CC(C)(C)C1=NC(=CC=C1)C(C)(C)C | |
191.32 | |
191.32 | |
Liquid |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee
GHS Signal Word: Warning
EINECSNumber : 209-557-6
RUO â Research Use Only