missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,6-Di-tert-butylpyridine, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC204640250

 View more versions of this product

Catalog No. AC204640250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

2, 6-Di-tert-butylpyridine
72°C
Glass bottle
0.852
Yellow
25g
585-48-8
100.0
C13H21N
MFCD00006306
01,212
2,6-di tert-butyl pyridine, 2,6-di-t-butyl pyridine, 2,6-di-t-butylpyridine, 2,6-di-tert-butyl pyridine, 2,6-di-tert-butylpyridine, oi9lf0h4mm, pubchem16119, pyridine, 2,6-bis 1,1-dimethylethyl, pyridine, 2,6-di-tert-butyl, unii-oi9lf0h4mm
UWKQJZCTQGMHKD-UHFFFAOYSA-N
2,6-ditert-butylpyridine
68510
Liquid
0.8520g/mL
Authentic
1.4720 to 1.4740
208.0°C
2.0°C to 4.0°C
97%
96.0
96% min. (GC)
(C5H3N[C(CH3)3]2
20, III, 2868
15, 3045
Solubility in water: insoluble. Other solubilities: soluble in organic solvents
CC(C)(C)C1=NC(=CC=C1)C(C)(C)C
191.32
191.32
97%

Chemical Identifiers

585-48-8
191.32
UWKQJZCTQGMHKD-UHFFFAOYSA-N
68510
CC(C)(C)C1=NC(=CC=C1)C(C)(C)C
C13H21N
MFCD00006306
2,6-di tert-butyl pyridine, 2,6-di-t-butyl pyridine, 2,6-di-t-butylpyridine, 2,6-di-tert-butyl pyridine, 2,6-di-tert-butylpyridine, oi9lf0h4mm, pubchem16119, pyridine, 2,6-bis 1,1-dimethylethyl, pyridine, 2,6-di-tert-butyl, unii-oi9lf0h4mm
2,6-ditert-butylpyridine
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee

GHS Signal Word: Warning

EINECSNumber : 209-557-6

SDS Documents