Learn More
2,6-Di-tert-butylpyridine, 97+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H5576103
Description
2,6-Di-tert-butylpyridine is used in the preparation of 2, 6-di-tert-butylpyridine hydrotriflate. It is used as a proton scavenger to check the progress of the living polymerization of isobutylene. It is associated with cerric ammonium nitrate and used in the alfa-enolation of aldehydes. It is involved in the preparation of vinyl triflate using polymer-bound 2,6-di-tert-butylpyridine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,6-Di-tert-butylpyridine is used in the preparation of 2, 6-di-tert-butylpyridine hydrotriflate. It is used as a proton scavenger to check the progress of the living polymerization of isobutylene. It is associated with cerric ammonium nitrate and used in the alfa-enolation of aldehydes. It is involved in the preparation of vinyl triflate using polymer-bound 2,6-di-tert-butylpyridine.
Solubility
Miscible with alcohol, acetone, and hexane. Immiscible with water.
Notes
Store in a cool place. Incompatible with strong acids and strong oxidizing agents.
Specifications
2,6-Di-tert-butylpyridine | |
0.867 | |
71°C (160°F) | |
1.473 | |
1 g | |
14,3036 | |
Miscible with alcohol,acetone,and hexane. Immiscible with water. | |
CC(C)(C)C1=NC(=CC=C1)C(C)(C)C | |
191.318 | |
191.32 |
585-48-8 | |
100°C to 101°C (23 mmHg) | |
C13H21N | |
MFCD00006306 | |
125886 | |
2,6-di-tert-butylpyridine, 2,6-di tert-butyl pyridine, 2,6-di-t-butylpyridine, unii-oi9lf0h4mm, 2,6-di-t-butyl pyridine, pyridine, 2,6-bis 1,1-dimethylethyl, oi9lf0h4mm, 2,6-di-tert-butyl pyridine, pyridine, 2,6-di-tert-butyl, pubchem16119 | |
UWKQJZCTQGMHKD-UHFFFAOYSA-N | |
2,6-ditert-butylpyridine | |
68510 | |
≥97% |
Chemical Identifiers
585-48-8 | |
191.318 | |
UWKQJZCTQGMHKD-UHFFFAOYSA-N | |
68510 | |
CC(C)(C)C1=NC(=CC=C1)C(C)(C)C |
C13H21N | |
MFCD00006306 | |
2,6-di-tert-butylpyridine, 2,6-di tert-butyl pyridine, 2,6-di-t-butylpyridine, unii-oi9lf0h4mm, 2,6-di-t-butyl pyridine, pyridine, 2,6-bis 1,1-dimethylethyl, oi9lf0h4mm, 2,6-di-tert-butyl pyridine, pyridine, 2,6-di-tert-butyl, pubchem16119 | |
2,6-ditert-butylpyridine |
Safety and Handling
GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H227-H315-H319-H335
EINECSNumber : 209-557-6
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only