missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,6-Di-tert-butylphenol, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC113002500

 View more versions of this product

Catalog No. AC113002500



Description

Description

Specifications

Specifications

2, 6-Di-tert-butylphenol
Authentic
0.1% max. (K.F.)
White to Yellow
250g
98.5
99%
[(CH3)3C]2C6H3OH
06, III, 2061
Solubility in water: insoluble. Other solubilities: soluble in alcohol and benzene
CC(C)(C)C1=CC=CC(=C1O)C(C)(C)C
206.33
206.33
99%
118°C
Glass bottle
253°C
35°C to 38°C
128-39-2
100.0
C14H22O
MFCD00008820
2,6 di-tert-butylphenol, 2,6-bis 1,1-dimethylethyl phenol, 2,6-bis tert-butyl phenol, 2,6-di-t-butylphenol, 2,6-di-tert-butylphenol, ethanox 701, ethyl 701, ethyl an 701, isonox 103, phenol, 2,6-bis 1,1-dimethylethyl
DKCPKDPYUFEZCP-UHFFFAOYSA-N
2,6-ditert-butylphenol
31405
Crystalline Solid

Chemical Identifiers

128-39-2
206.33
DKCPKDPYUFEZCP-UHFFFAOYSA-N
31405
CC(C)(C)C1=CC=CC(=C1O)C(C)(C)C
C14H22O
MFCD00008820
2,6 di-tert-butylphenol, 2,6-bis 1,1-dimethylethyl phenol, 2,6-bis tert-butyl phenol, 2,6-di-t-butylphenol, 2,6-di-tert-butylphenol, ethanox 701, ethyl 701, ethyl an 701, isonox 103, phenol, 2,6-bis 1,1-dimethylethyl
2,6-ditert-butylphenol
Safety and Handling

Safety and Handling

GHS H Statement
Toxic to aquatic life with long lasting effects.
Harmful if swallowed.

GHS P Statement
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

GHS Signal Word: Warning

EINECSNumber : 204-884-

SDS Documents
Product Certifications