Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,6-Di-tert-butyl-p-benzoquinone-4-oxime, 98+%


Quantity 5g

Manufacturer: alfa aesar™  L09903-06

 View more versions of this product

Catalog No. AAL0990306

  • / Each
 

Description & Specifications

Chemical Identifiers

Melting Point 218° to 220°C
Quantity 5g
Chemical Name or Material 2,6-Di-tert-butyl-p-benzoquinone-4-oxime
CAS 15052-28-5
Assay ≥98%
Molecular Formula C14H21NO2
Formula Weight 235.33g/mol
MDL Number MFCD00135757
Beilstein 1966406
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Specifications

Chemical Name or Material 2,6-Di-tert-butyl-p-benzoquinone-4-oxime
CAS 15052-28-5
Molecular Formula C14H21NO2
Formula Weight 235.33g/mol
MDL Number MFCD00135757
Beilstein 1966406
Accessories

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.