missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,6-Di-tert-butyl-p-benzoquinone-4-oxime, 98+%

Manufacturer:  Alfa Aesar™ L0990314

 View more versions of this product

Catalog No. AAL0990314



Specifications

Specifications

2,6-Di-tert-butyl-p-benzoquinone-4-oxime
25g
C14H21NO2
1966406
JLOKPJOOVQELMO-UHFFFAOYSA-N
2,6-ditert-butyl-4-nitrosophenol
70383
≥98%
218°C to 220°C
15052-28-5
MFCD00135757
2,6-di-tert-butyl-4-nitrosophenol, phenol, 2,6-bis 1,1-dimethylethyl-4-nitroso, 2,6-di tert-butyl-4-nitrosophenol, phenol, 2,6-di-tert-butyl-4-nitroso, 2,6-di-tert-butyl-4-hydroxyimino cyclohexa-2,5-dienone, 2,6-di-tert-butyl-p-benzoquinone-4-oxime, 2,6-di-tert-butyl-4-hydroxyimino cyclohexa-2,5-dien-1-one, 2,6-di-t-butyl-4-nitrosophenol, 4-nitroso-2,6-di-t-butylphenol, fxyztgvhlfvvnn-uhfffaoysa-n
CC(C)(C)C1=CC(=CC(=C1O)C(C)(C)C)N=O
235.327
235.33

Chemical Identifiers

15052-28-5
235.327
JLOKPJOOVQELMO-UHFFFAOYSA-N
70383
CC(C)(C)C1=CC(=CC(=C1O)C(C)(C)C)N=O
C14H21NO2
MFCD00135757
2,6-di-tert-butyl-4-nitrosophenol, phenol, 2,6-bis 1,1-dimethylethyl-4-nitroso, 2,6-di tert-butyl-4-nitrosophenol, phenol, 2,6-di-tert-butyl-4-nitroso, 2,6-di-tert-butyl-4-hydroxyimino cyclohexa-2,5-dienone, 2,6-di-tert-butyl-p-benzoquinone-4-oxime, 2,6-di-tert-butyl-4-hydroxyimino cyclohexa-2,5-dien-1-one, 2,6-di-t-butyl-4-nitrosophenol, 4-nitroso-2,6-di-t-butylphenol, fxyztgvhlfvvnn-uhfffaoysa-n
2,6-ditert-butyl-4-nitrosophenol
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents