Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,6-Di-tert-butyl-4-methylpyrylium trifluoromethanesulfonate, 95%

$33.15 - $94.93

Chemical Identifiers

Chemical Name or Material 2,6-Di-tert-butyl-4-methylpyrylium trifluoromethanesulfonate
CAS 59643-43-5
Molecular Formula C15H23F3O4S
Formula Weight 356.41
MDL Number MFCD00229854
Beilstein 3644038
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1630403

  SDS
alfa aesar™
L1630403
1g Each for $33.15

AAL1630406

  SDS
alfa aesar™
L1630406
5g Each for $94.93
Description & Specifications

Specifications

Formula Weight 356.41
Chemical Name or Material 2,6-Di-tert-butyl-4-methylpyrylium trifluoromethanesulfonate
CAS 59643-43-5
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 95%
Health Hazard 1 Warning
Molecular Formula C15H23F3O4S
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Beilstein 3644038
MDL Number MFCD00229854
Quantity 1g
Melting Point ≈163°C
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.