Please login to your online account to display your discounted pricing

2,6-Di-tert-butyl-4-methylpyridine, 98%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC224520050

 View more versions of this product

Catalog No. AC224520050

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Liquid or Low Melting Solid
Formula Weight 205.34
Quantity 5g
Color Colorless to yellow
Fieser 09,141; 10,123; 11,160; 16,110
Merck Index 15, 3042
CAS Max % 100.0
Beilstein 20, V,6, 190
Chemical Name or Material 2, 6-Di-tert-butyl-4-methylpyridine
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4760 to 1.4780 (20°C, 589nm)
Assay Percent Range 98%
Molecular Formula C14H23N
Linear Formula {(CH3)3C}2C5H2NCH3
MDL Number MFCD00006305
CAS 38222-83-2
CAS Min % 97.5
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 2, 6-Di-tert-butyl-4-methylpyridine
CAS 38222-83-2
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C14H23N
Linear Formula {(CH3)3C}2C5H2NCH3
Formula Weight 205.34
MDL Number MFCD00006305
Accessories

GHS Signal Word: Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

SDS & Safety Documents