Please login to your online account to display your discounted pricing

2,6-Di-tert-butyl-4-methoxyphenol, 97%, ACROS Organics™

$47.19 - $131.31

Chemical Identifiers

Chemical Name or Material 2, 6-Di-tert-butyl-4-methoxyphenol
Name Note 97%
CAS 489-01-0
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0% (GC)
Assay Percent Range 97%
Molecular Formula C15H24O2
Formula Weight 236.35
MDL Number MFCD00008824
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC294160250

  SDS
fisher scientific acros
294160250
Glass bottle 25g Each for $47.19

AC294161000

  SDS
thermo fisher scientific
294161000
Glass bottle 100g Each for $131.31
Description & Specifications

Specifications

Physical Form Liquid
Assay ≥96.0% (GC)
Formula Weight 236.35
Melting Point 102° to 106°C
Quantity 25g
Color Clear
EINECS Number 207-693-0
TSCA TSCA
RTECS Number SJ7785668
Fieser 16,110; 16,58; 16,58
CAS Min % 96.0
CAS Max % 100.0
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Chemical Name or Material 2, 6-Di-tert-butyl-4-methoxyphenol
Infrared Spectrum Authentic
Packaging Glass bottle
Name Note 97%
Assay Percent Range 97%
Molecular Formula C15H24O2
MDL Number MFCD00008824
CAS 489-01-0
Synonym 2, 6-Bis(1, 1-dimethylethyl)-4-methoxyphenol
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.