missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,6-Di-tert-butyl-4-ethylphenol, 98%

$44.50 - $146.85

Chemical Identifiers

CAS 4130-42-1
Molecular Formula C16H26O
Formula Weight 234.38
MDL Number MFCD00017366
Beilstein 1875693
Chemical Name or Material 2,6-Di-tert-butyl-4-ethylphenol
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0868330 SDS Alfa Aesar™
L0868330
250g Each for $146.85
AAL0868318 SDS Alfa Aesar™
L0868318
50g Each for $44.50
Specifications Description & Specifications

Chemical Identifiers

4130-42-1
234.38
1875693
C16H26O
MFCD00017366
2,6-Di-tert-butyl-4-ethylphenol

Specifications

4130-42-1
272°C
105°C (221°F)
234.38
1875693
98%
42° to 44°C
250g
C16H26O
MFCD00017366
2,6-Di-tert-butyl-4-ethylphenol

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA