missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(6-Chloro-3-pyridazinylamino)ethanol, 97%

$69.58 - $834.14

Chemical Identifiers

CAS 51947-89-8
Molecular Formula C6H8ClN3O
Molecular Weight (g/mol) 173.6
InChI Key OCSNMMYFHUBJGA-UHFFFAOYSA-N
Synonym 2-6-chloro-3-pyridazinyl amino ethanol
PubChem CID 582312
IUPAC Name 2-[(6-chloropyridazin-3-yl)amino]ethanol
SMILES C1=CC(=NN=C1NCCO)Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH61432MD View Documents Alfa Aesar™
H61432MD
250mg Each for $69.58
 
AAH6143203 View Documents Alfa Aesar™
H6143203
1g Each for $207.32
 
AAH6143206 View Documents Alfa Aesar™
H6143206
5g Each for $834.14
 
Specifications

Chemical Identifiers

51947-89-8
173.6
2-6-chloro-3-pyridazinyl amino ethanol
2-[(6-chloropyridazin-3-yl)amino]ethanol
C6H8ClN3O
OCSNMMYFHUBJGA-UHFFFAOYSA-N
582312
C1=CC(=NN=C1NCCO)Cl

Specifications

250mg
C6H8ClN3O
OCSNMMYFHUBJGA-UHFFFAOYSA-N
2-[(6-chloropyridazin-3-yl)amino]ethanol
582312
Solid
2-(6-Chloro-3-pyridazinylamino)ethanol
51947-89-8
2-6-chloro-3-pyridazinyl amino ethanol
C1=CC(=NN=C1NCCO)Cl
173.6
173.6
97%