Please login to your online account to display your discounted pricing

2-(6-Bromonaphthyl)-β-D-xylopyranoside hydrate, Acros Organics


Quantity 100mg
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC341151000

 View more versions of this product

Catalog No. AC341151000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Powder
Formula Weight 355.19
Quantity 100mg
Color White to off-white
Water 5% Max. (K.F.)
Melting Point 190° to 196°C
Assay Passes Test
Molecular Formula C15H15BrO5·xH2O
CAS 69594-75-8
Appearance of Solution (1% acetone/water 1:1) clear, colorless
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material 2-(6-Bromonaphthyl)-β-D-xylopyranoside hydrate
MDL Number MFCD04039743
Recommended Storage Refrigerator +4°C

Specifications

Chemical Name or Material 2-(6-Bromonaphthyl)-β-D-xylopyranoside hydrate
Assay Passes Test
Molecular Formula C15H15BrO5·xH2O
Formula Weight 355.19
MDL Number MFCD04039743
Accessories

Refrigerator +4°C

SDS & Safety Documents