Please login to your online account to display your discounted pricing

2-(6-Bromonaphthyl)-β-D-xylopyranoside hydrate, Acros Organics

Chemical Identifiers

Chemical Name or Material 2-(6-Bromonaphthyl)-β-D-xylopyranoside hydrate
Assay Passes Test
Molecular Formula C15H15BrO5·xH2O
Formula Weight 355.19
MDL Number MFCD04039743
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC341151000

  SDS
thermo fisher scientific
AC341151000
Glass bottle 100mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC341155000

  SDS
thermo fisher scientific
AC341155000
Glass bottle 500mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 355.19
Physical Form Powder
Quantity 100mg
Color White to off-white
Assay Passes Test
Water 5% Max. (K.F.)
Melting Point 190° to 196°C
Infrared Spectrum Authentic
CAS 69594-75-8
Packaging Glass bottle
Appearance of Solution (1% acetone/water 1:1) clear, colorless
Molecular Formula C15H15BrO5·xH2O
Recommended Storage Refrigerator +4°C
Chemical Name or Material 2-(6-Bromonaphthyl)-β-D-xylopyranoside hydrate
MDL Number MFCD04039743
Safety and Handling

Refrigerator +4°C