missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,6-Bis(dodecyloxy)benzaldehyde, 97%

$38.55 - $453.38

Chemical Identifiers

CAS 1198467-57-0
Molecular Formula C31H54O3
Molecular Weight (g/mol) 474.77
MDL Number MFCD25372091
InChI Key QPAKGKFSBBAHMS-UHFFFAOYSA-N
Synonym 2,6-bis dodecyloxy benzaldehyde, 2,6-di dodecyloxy benzaldehyde, 2,6-bis dodecyloxy benzaldehyde
PubChem CID 75201262
IUPAC Name 2,6-didodecoxybenzaldehyde
SMILES CCCCCCCCCCCCOC1=C(C(=CC=C1)OCCCCCCCCCCCC)C=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH64087MD View Documents Alfa Aesar™
H64087MD
250mg Each for $38.55
 
AAH6408703 View Documents Alfa Aesar™
H6408703
1g Each for $113.97
 
AAH6408706 View Documents Alfa Aesar™
H6408706
5g Each for $453.38
 
Specifications

Chemical Identifiers

1198467-57-0
474.77
QPAKGKFSBBAHMS-UHFFFAOYSA-N
75201262
CCCCCCCCCCCCOC1=C(C(=CC=C1)OCCCCCCCCCCCC)C=O
C31H54O3
MFCD25372091
2,6-bis dodecyloxy benzaldehyde, 2,6-di dodecyloxy benzaldehyde, 2,6-bis dodecyloxy benzaldehyde
2,6-didodecoxybenzaldehyde

Specifications

Yellow
1198467-57-0
MFCD25372091
2,6-bis dodecyloxy benzaldehyde, 2,6-di dodecyloxy benzaldehyde, 2,6-bis dodecyloxy benzaldehyde
CCCCCCCCCCCCOC1=C(C(=CC=C1)OCCCCCCCCCCCC)C=O
474.77
474.77
97%
250mg
C31H54O3
20848961
QPAKGKFSBBAHMS-UHFFFAOYSA-N
2,6-didodecoxybenzaldehyde
75201262
Solid
2,6-Bis(dodecyloxy)benzaldehyde