missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,6-Bis(dodecyloxy)benzaldehyde, 97%

Manufacturer:  Alfa Aesar™ H6408706

 View more versions of this product

Catalog No. AAH6408706



Specifications

Specifications

2,6-Bis(dodecyloxy)benzaldehyde
5g
C31H54O3
20848961
QPAKGKFSBBAHMS-UHFFFAOYSA-N
2,6-didodecoxybenzaldehyde
75201262
Solid
Yellow
1198467-57-0
MFCD25372091
2,6-bis dodecyloxy benzaldehyde, 2,6-di dodecyloxy benzaldehyde, 2,6-bis dodecyloxy benzaldehyde
CCCCCCCCCCCCOC1=C(C(=CC=C1)OCCCCCCCCCCCC)C=O
474.77
474.77
97%

Chemical Identifiers

1198467-57-0
474.77
QPAKGKFSBBAHMS-UHFFFAOYSA-N
75201262
CCCCCCCCCCCCOC1=C(C(=CC=C1)OCCCCCCCCCCCC)C=O
C31H54O3
MFCD25372091
2,6-bis dodecyloxy benzaldehyde, 2,6-di dodecyloxy benzaldehyde, 2,6-bis dodecyloxy benzaldehyde
2,6-didodecoxybenzaldehyde
SDS Documents