missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,6-Bis(4-fluorobenzylidene)cyclohexanone, TCI America™

$172.64 - $525.56

Chemical Identifiers

CAS 62085-74-9
Molecular Formula C20H16F2O
Molecular Weight (g/mol) 310.344
MDL Number MFCD00112608
InChI Key STPNBQOTKFPYEG-UNZYHPAISA-N
Synonym 2,6-Bis(4-fluorobenzal)cyclohexanone
PubChem CID 1551384
IUPAC Name (2E,6E)-2,6-bis[(4-fluorophenyl)methylidene]cyclohexan-1-one
SMILES C1CC(=CC2=CC=C(C=C2)F)C(=O)C(=CC3=CC=C(C=C3)F)C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B22825G View Documents TCI America
B22825G
5g Each for $172.64
 
B228225G View Documents TCI America
B228225G
25g Each for $525.56
 
Specifications

Chemical Identifiers

62085-74-9
310.344
STPNBQOTKFPYEG-UNZYHPAISA-N
1551384
C1CC(=CC2=CC=C(C=C2)F)C(=O)C(=CC3=CC=C(C=C3)F)C1
C20H16F2O
MFCD00112608
2,6-Bis(4-fluorobenzal)cyclohexanone
(2E,6E)-2,6-bis[(4-fluorophenyl)methylidene]cyclohexan-1-one

Specifications

Yellow
62085-74-9
MFCD00112608
STPNBQOTKFPYEG-UNZYHPAISA-N
(2E,6E)-2,6-bis[(4-fluorophenyl)methylidene]cyclohexan-1-one
1551384
Crystal-Powder at 20°C
2,6-Bis(4-fluorobenzylidene)cyclohexanone
5g
C20H16F2O
2,6-Bis(4-fluorobenzal)cyclohexanone
C1CC(=CC2=CC=C(C=C2)F)C(=O)C(=CC3=CC=C(C=C3)F)C1
310.344
310.34
≥96.0% (HPLC)
Safety and Handling

Safety and Handling

TSCA : No