Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,6,8-Trimethyl-4-nonanone, 98%

$115.45 - $469.49

Chemical Identifiers

Chemical Name or Material 2,6,8-Trimethyl-4-nonanone
CAS 123-18-2
Molecular Formula C12H22O
Formula Weight 182.31
MDL Number MFCD00048361
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2051809

 
alfa aesar™
B2051809
10g Each for $115.45

AAB2051818

 
alfa aesar™
B2051818
50g Each for $469.49
Description & Specifications

Specifications

Formula Weight 182.31
Chemical Name or Material 2,6,8-Trimethyl-4-nonanone
TSCA TSCA
Assay 98%
Molecular Formula C12H22O
MDL Number MFCD00048361
CAS 123-18-2
Quantity 10g

Applications
The primary uses of 2,6,8-Trimethyl-4-nonanone, are as a cleaning and degreasing solvent, an extraction solvent in mining, a chemical intermediate to produce 2,6,8-trimethyl-4-nonanol,and a solvent for certain coatings. It is also used as a chemical intermediate - to produce 2,6,8-trimethyl-4-nonanol. It is used in inks, fragrance as a solvent.

Solubility
Soluble in alcohol and insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.