Learn More
2(5H)-Furanone, 96%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L1314214
Description
2(5H)-Furanone is employed in the preparation of 5-substituted 2(5H) furanones (gamma-butenolides) catalyzed by bifunctional aminothiourea and aminosquaramide organocatalysts through direct aldol reaction with aromatic aldehydes. Further, it is used in the synthesis of lignans by Michael addition reactions. In addition to this, it is used in three-component Michael-Aldol reactions with an aldehyde and thiolate or carbanion.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2(5H)-Furanone is employed in the preparation of 5-substituted 2(5H) furanones (gamma-butenolides) catalyzed by bifunctional aminothiourea and aminosquaramide organocatalysts through direct aldol reaction with aromatic aldehydes. Further, it is used in the synthesis of lignans by Michael addition reactions. In addition to this, it is used in three-component Michael-Aldol reactions with an aldehyde and thiolate or carbanion.
Solubility
Immiscible with water.
Notes
Light and moisture sensitive. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
Specifications
2(5H)-Furanone | |
3°C to 4°C | |
212°C to 214°C | |
C4H4O2 | |
MFCD00005376 | |
383585 | |
2 5h-furanone, furan-2 5h-one, butenolide, 2-butenolide, gamma-crotonolactone, 2-buten-4-olide, isocrotonolactone, 2-oxo-2,5-dihydrofuran, crotonolactone, 2-5h-furanone | |
VIHAEDVKXSOUAT-UHFFFAOYSA-N | |
2H-furan-5-one | |
10341 | |
84.07 |
497-23-4 | |
1.197 | |
101°C (213°F) | |
1.469 | |
25 g | |
Light sensitive | |
Immiscible with water. | |
O=C1OCC=C1 | |
84.07 | |
CHEBI:38118 | |
98% |
Chemical Identifiers
497-23-4 | |
84.07 | |
VIHAEDVKXSOUAT-UHFFFAOYSA-N | |
10341 | |
2H-furan-5-one |
C4H4O2 | |
MFCD00005376 | |
2 5h-furanone, furan-2 5h-one, butenolide, 2-butenolide, gamma-crotonolactone, 2-buten-4-olide, isocrotonolactone, 2-oxo-2,5-dihydrofuran, crotonolactone, 2-5h-furanone | |
CHEBI:38118 | |
O=C1OCC=C1 |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 207-839-3
RTECSNumber : LU3453000
TSCA : No
Recommended Storage : Keep cold; Store under Nitrogen
RUO – Research Use Only